CAS RN 149184-21-4
IUPAC Name:
hydrogen (cyclohexylmethyl)[(2S)-3-{[(1S)-1-(3,4-dichlorophenyl)ethyl]amino}-2-hydroxypropyl]phosphinic acid chloride
Synonyms:
hydrogen cyclohexylmethyl((2S)-3-{[(1S)-1-(3,4-dichlorophenyl)ethyl]amino}-2-hydroxypropyl)phosphinic acid chloride
Molecular Weight (g/mol):
444.76
Molecular Formula:
C18H29Cl3NO3P
InChi Key:
ZQCFHOVIXCJPLE-LINSIKMZSA-N
SMILES:
[H+].[Cl-].C[C@H](NC[C@H](O)CP(O)(=O)CC1CCCCC1)C1=CC=C(Cl)C(Cl)=C1
1
–
1
of
1
results
1
Tocris Bioscience™ CGP 54626 hydrochloride
CAS: 149184-21-4 Molecular Formula: C18H29Cl3NO3P