accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3ClClHNPOHOHOHCl

CAS RN 149184-22-5

IUPAC Name: benzyl(3-{[1-(3,4-dichlorophenyl)ethyl]amino}-2-hydroxypropyl)phosphinic acid hydrochloride
Synonyms: benzyl(3-{[1-(3,4-dichlorophenyl)ethyl]amino}-2-hydroxypropyl)phosphinic acid hydrochloride
Molecular Weight (g/mol): 438.71
Molecular Formula: C18H23Cl3NO3P
InChi Key: PXQAIXBYWZBYKJ-UHFFFAOYNA-N
SMILES: Cl.CC(NCC(O)CP(O)(=O)CC1=CC=CC=C1)C1=CC(Cl)=C(Cl)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results