accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COO

CAS RN 14924-53-9

IUPAC Name: ethyl cyclobutanecarboxylate
Synonyms: cyclobutanecarboxylic acid ethyl ester
Molecular Weight (g/mol): 128.17
Molecular Formula: C7H12O2
InChi Key: SMVBADCAMQOTOV-UHFFFAOYSA-N
SMILES: CCOC(=O)C1CCC1
Boiling Point 
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results