accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClH3CNH3CNNOONH3CCH3NH3CCH3O(E/Z)(?)

CAS RN 14969-56-3

IUPAC Name: 3,6-bis(diethylamino)-9-(2-{[8-(diethylamino)-12H-10-oxa-5-azatetraphen-12-ylidene]carbamoyl}phenyl)-10λ⁴-xanthen-10-ylium chloride
Synonyms: 3,6-bis(diethylamino)-9-(2-{[8-(diethylamino)-10-oxa-5-azatetraphen-12-ylidene]carbamoyl}phenyl)-10λ⁴-xanthen-10-ylium chloride
Molecular Weight (g/mol): 778.39
Molecular Formula: C48H48ClN5O3
InChi Key: UYQVPCFMVWEDCA-UHFFFAOYSA-M
SMILES: [Cl-].CCN(CC)C1=CC2=[O+]C3=CC(=CC=C3C(C3=CC=CC=C3C(=O)N=C3C=C4OC5=CC(=CC=C5N=C4C4=CC=CC=C34)N(CC)CC)=C2C=C1)N(CC)CC
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results