CAS RN 149719-06-2
IUPAC Name:
hydrogen 3-(piperidin-1-yl)propyl 4-amino-5-chloro-2-methoxybenzoate chloride
Synonyms:
hydrogen 3-(piperidin-1-yl)propyl 4-amino-5-chloro-2-methoxybenzoate chloride
Molecular Weight (g/mol):
363.28
Molecular Formula:
C16H24Cl2N2O3
InChi Key:
QLZLBYYNMGQIAR-UHFFFAOYSA-N
SMILES:
[H+].[Cl-].COC1=CC(N)=C(Cl)C=C1C(=O)OCCCN1CCCCC1
1
–
1
of
1
results
1
R&D Systems™ RS 23597-190 hydrochloride
CAS: 149719-06-2 Molecular Formula: C16H24Cl2N2O3