CAS RN 14985-44-5
IUPAC Name:
(2R,3S,5R)-5-(6-amino-8-bromo-9H-purin-9-yl)-2-(hydroxymethyl)oxolan-3-ol
Synonyms:
(2R,3S,5R)-5-(6-amino-8-bromopurin-9-yl)-2-(hydroxymethyl)oxolan-3-ol
Molecular Weight (g/mol):
330.14
Molecular Formula:
C10H12BrN5O3
InChi Key:
NJBIVXMQFIQOGE-DWFKHXNSNA-N
SMILES:
NC1=C2N=C(Br)N([C@H]3C[C@H](O)[C@@H](CO)O3)C2=NC=N1
1
–
1
of
1
results
1
8-Bromo-2'-deoxyadenosine, 99%, Thermo Scientific Chemicals
CAS: 14985-44-5 Molecular Formula: C10H12BrN5O3 Molecular Weight (g/mol): 330.14 MDL Number: MFCD01630970 InChI Key: NJBIVXMQFIQOGE-DWFKHXNSNA-N Synonym: 8-bromo-2'-deoxyadenosine,2r,3s,5r-5-6-amino-8-bromo-9h-purin-9-yl-2-hydroxymethyl tetrahydrofuran-3-ol,2r,3s,5r-5-6-amino-8-bromopurin-9-yl-2-hydroxymethyl oxolan-3-ol,adenosine,8-bromo-2'-deoxy,2'-deoxy-8-bromoadenosine,8-b-2-da,8-bromo-2?-deoxyadenosine,adenosine, 8-bromo-2'-deoxy,8-bromo-2-deoxyadenosine PubChem CID: 96547 SMILES: NC1=C2N=C(Br)N([C@H]3C[C@H](O)[C@@H](CO)O3)C2=NC=N1