accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 15016-43-0

IUPAC Name: (3-ethenylphenyl)boronic acid
Synonyms: 3-ethenylphenylboronic acid
Molecular Weight (g/mol): 147.97
Molecular Formula: C8H9BO2
InChi Key: SYBQEKBVWDPVJM-UHFFFAOYSA-N
SMILES: OB(O)C1=CC=CC(C=C)=C1
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)