Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Vinylbenzeneboronic acid, 98%, Thermo Scientific™

Catalog No. AAL19829MD
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL19829MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL19829MD Supplier Thermo Scientific Chemicals Supplier No. L19829MD

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15016-43-0
Molecular Formula C8H9BO2
Molecular Weight (g/mol) 147.968
MDL Number MFCD01074679
InChI Key SYBQEKBVWDPVJM-UHFFFAOYSA-N
Synonym 3-vinylphenylboronic acid, 3-vinylphenyl boronic acid, 3-vinylbenzeneboronic acid, styrene-3-boronic acid, 3-ethenylphenyl boronic acid, m-vinylbenzeneboronic acid, boronic acid, 3-ethenylphenyl, 3-vinylphenylbronicacid, 3-vinyl-phenylboronic acid
PubChem CID 4366886
IUPAC Name (3-ethenylphenyl)boronic acid
SMILES B(C1=CC(=CC=C1)C=C)(O)O

Specifications

Melting Point 127°C to 129°C
Quantity 250 mg
Beilstein 3030426
Formula Weight 147.97
Percent Purity 98%
Chemical Name or Material 3-Vinylbenzeneboronic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Keep cold
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.