CAS RN 150323-78-7
IUPAC Name:
2-(piperazin-1-yl)quinoline; bis(but-2-enedioic acid)
Synonyms:
bis(butenedioic acid); quipazine
Molecular Weight (g/mol):
445.43
Molecular Formula:
C21H23N3O8
InChi Key:
VAOSOCRJSSWBEQ-UHFFFAOYSA-N
SMILES:
OC(=O)C=CC(O)=O.OC(=O)C=CC(O)=O.C1CN(CCN1)C1=CC=C2C=CC=CC2=N1
1
–
1
of
1
results
1
R&D Systems™ Quipazine dimaleate
CAS: 150323-78-7 Molecular Formula: C21H23N3O8