CAS RN 150333-69-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
MilliporeSigma™ Calbiochem™ Deoxyhypusine Synthase Inhibitor, GC7
The Deoxyhypusine Synthase Inhibitor, GC7, also referenced under CAS 150333-69-0, controls the biological activity of Deoxyhypusine Synthase.