CAS RN 150374-95-1
IUPAC Name:
sodium 2-[(2-{4-[(2,2-dimethylpropanoyl)oxy]benzenesulfonamido}phenyl)formamido]acetate
Synonyms:
sodium 2-[(2-{4-[(2,2-dimethylpropanoyl)oxy]benzenesulfonamido}phenyl)formamido]acetate
Molecular Weight (g/mol):
456.44
Molecular Formula:
C20H21N2NaO7S
InChi Key:
ZAIFANJZUGNYCK-UHFFFAOYSA-M
SMILES:
[Na+].CC(C)(C)C(=O)OC1=CC=C(C=C1)S(=O)(=O)NC1=CC=CC=C1C(=O)NCC([O-])=O
1
–
1
of
1
results
1
R&D Systems™ Sivelestat sodium salt
CAS: 150374-95-1 Molecular Formula: C20H21N2NaO7S