accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1505-50-6

IUPAC Name: 3-(4-methylphenyl)propanoic acid
Synonyms: 3-(P-tolyl)propionic acid
Molecular Weight (g/mol): 164.2
Molecular Formula: C10H12O2
InChi Key: LDYGRLNSOKABMM-UHFFFAOYSA-N
SMILES: CC1=CC=C(CCC(O)=O)C=C1
Percent Purity 
  • (2)
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)