Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-(4-Methylphenyl)propionic acid, 98+%, Thermo Scientific™

Catalog No. AAA1451309
Change view
Click to view available options
Quantity:
5 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1451309 10 g
AAA1451306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1451309 Supplier Thermo Scientific Chemicals Supplier No. A1451309

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
3-(p-Tolyl)propionic acid was used during the coupling of 1-(3-dimethylaminopropyl)-3-ethylcarbodiimide with 4,5-diphenyl-2-(2-p-tolyl-ethyl)-4,5-dihydro-1H-imidazole resin.

Solubility
Soluble in methanol.

Notes
Keep container tightly closed. Store in a cool and dark place. Store away from incompatible materials such as oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1505-50-6
Molecular Formula C10H12O2
Molecular Weight (g/mol) 164.2
MDL Number MFCD00016561
InChI Key LDYGRLNSOKABMM-UHFFFAOYSA-N
Synonym 3-4-methylphenyl propionic acid, 3-p-tolyl propionic acid, 3-p-tolyl propanoic acid, benzenepropanoic acid, 4-methyl, 3-4-methylphenyl propanoic acid, 4-methylphenylpropionic acid, 3-p-tolylpropanoic acid, p-methylhydrocinnamic acid, 4-methyl phenyl propionic acid, 4-methylhydrocinnamic acid
PubChem CID 73927
IUPAC Name 3-(4-methylphenyl)propanoic acid
SMILES CC1=CC=C(CCC(O)=O)C=C1

Specifications

Melting Point 115°C to 117°C
Quantity 10 g
Beilstein 2046164
Solubility Information Soluble in methanol.
Formula Weight 164.2
Percent Purity ≥98%
Chemical Name or Material 3-(4-Methylphenyl)propionic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 216-132-9
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.