Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-(p-Tolyl)propionic Acid 98.0+%, TCI America™
SDP

Catalog No. T27975G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T27975G 5 g
T279725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T27975G Supplier TCI America Supplier No. T27975G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1505-50-6
Molecular Formula C10H12O2
Molecular Weight (g/mol) 164.2
MDL Number MFCD00016561
InChI Key LDYGRLNSOKABMM-UHFFFAOYSA-N
Synonym 3-4-methylphenyl propionic acid, 3-p-tolyl propionic acid, 3-p-tolyl propanoic acid, benzenepropanoic acid, 4-methyl, 3-4-methylphenyl propanoic acid, 4-methylphenylpropionic acid, 3-p-tolylpropanoic acid, p-methylhydrocinnamic acid, 4-methyl phenyl propionic acid, 4-methylhydrocinnamic acid
PubChem CID 73927
IUPAC Name 3-(4-methylphenyl)propanoic acid
SMILES CC1=CC=C(CCC(O)=O)C=C1

Specifications

Melting Point 116°C
Color White-Yellow
Quantity 5 g
Formula Weight 164.2
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-(p-Tolyl)propionic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.