accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOOCH3

CAS RN 15057-11-1

IUPAC Name: 4-methoxybutyl acetate
Synonyms: 4-methoxybutyl acetate
Molecular Weight (g/mol): 146.19
Molecular Formula: C7H14O3
InChi Key: LMLBDDCTBHGHEO-UHFFFAOYSA-N
SMILES: COCCCCOC(C)=O
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results