Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Methoxybutyl Acetate 98.0+%, TCI America™
SDP

Catalog No. A16765G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A16765G 5 g
A16761G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A16765G Supplier TCI America Supplier No. A16765G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 15057-11-1
Molecular Formula C7H14O3
Molecular Weight (g/mol) 146.19
MDL Number MFCD06797048
InChI Key LMLBDDCTBHGHEO-UHFFFAOYSA-N
Synonym Acetic Acid 4-Methoxybutyl Ester
PubChem CID 21903822
IUPAC Name 4-methoxybutyl acetate
SMILES COCCCCOC(C)=O

Specifications

Color Yellow
Boiling Point 71°C
Quantity 5 g
UN Number 3272
Formula Weight 146.19
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Methoxybutyl Acetate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.