accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOHNOOCH3

CAS RN 15069-79-1

IUPAC Name: 5-(acetyloxy)-1H-indol-6-yl acetate
Synonyms: 5-(acetyloxy)-1H-indol-6-yl acetate
Molecular Weight (g/mol): 233.22
Molecular Formula: C12H11NO4
InChi Key: NTOLUQGMBCPVOZ-UHFFFAOYSA-N
SMILES: CC(=O)OC1=C(OC(C)=O)C=C2C=CNC2=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results