accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NNOOH

CAS RN 1508-75-4

IUPAC Name: N-ethyl-3-hydroxy-2-phenyl-N-[(pyridin-4-yl)methyl]propanamide
Synonyms: tropicamide I-picamide tropimil (r)-isomer tropicamide mydral mydrum tropicamide, (R)-isomer mydriaticum tropicamide faure triaminic DM
Molecular Weight (g/mol): 284.36
Molecular Formula: C17H20N2O2
InChi Key: BGDKAVGWHJFAGW-UHFFFAOYNA-N
SMILES: CCN(CC1=CC=NC=C1)C(=O)C(CO)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (1)
  • (5)
  • (2)
Percent Purity 
  • (1)
  • (1)
  • (5)
Grade 
  • (1)
  • (5)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (5)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (5)
  • (2)
  • (1)
  • (1)
  • (5)
  • (1)
  • (5)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)