Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tropicamide, R&D Systems™
SDP

Catalog No. 0909100 Shop All Novus Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
09-091-00 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 09-091-00 Supplier R&D Systems Supplier No. 0909/100

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Selective M4 antagonist

Tropicamide is an M4 selective muscarinic receptor antagonist.

Chemical Identifiers

CAS 1508-75-4
Molecular Formula C17H20N2O2
Molecular Weight (g/mol) 284.359
InChI Key BGDKAVGWHJFAGW-UHFFFAOYSA-N
Synonym tropicamide, mydriacyl, mydriaticum, mydriafair, tropicacyl, minims tropicamide, bistropamide, epitromina, tropicamidum, mydrum
PubChem CID 5593
IUPAC Name N-ethyl-3-hydroxy-2-phenyl-N-(pyridin-4-ylmethyl)propanamide
SMILES CCN(CC1=CC=NC=C1)C(=O)C(CO)C2=CC=CC=C2

Specifications

Quantity 100 mg
Formula Weight 284.36
Percent Purity >98%
Product Line Tocris
Chemical Name or Material Tropicamide
Recommended Storage Store at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.