CAS RN 150810-68-7
CAS RN 150810-68-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Tocris Bioscience™ 5-TAMRA, NHS
Fluorescent amine-reactive dye for labeling proteins, peptides and oligonucleotides