Learn More
Tocris Bioscience™ 5-TAMRA, NHS
Description
Please see the protocol for further information on protein/antibody labeling and conjugation.
It is recommended to prepare stock solutions in DMSO.
Specifications
Specifications
| Chemical Name or Material | 2-[3-(Dimethylamino)-6-dimethylazaniumylidenexanthen-9-yl]-5-(2,5-dioxopyrrolidin-1-yl)oxycarbonylbenzoate |
| CAS | 150810-68-7 |
| Quantity | 1 mg |
| Target | Fluorescent Probes |
| Molecular Formula | C29H25N3O7 |
| Purity | 0.9 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.