CAS RN 1511-16-6
IUPAC Name:
4-[4-(4-chlorophenyl)-4-hydroxypiperidin-1-yl]-1-(4-fluorophenyl)butan-1-one hydrochloride
Synonyms:
haloperidol hydrochloride
Molecular Weight (g/mol):
412.33
Molecular Formula:
C21H24Cl2FNO2
InChi Key:
JMRYYMBDXNZQMH-UHFFFAOYSA-N
SMILES:
Cl.OC1(CCN(CCCC(=O)C2=CC=C(F)C=C2)CC1)C1=CC=C(Cl)C=C1
1
–
1
of
1
results
1
R&D Systems™ Haloperidol hydrochloride
CAS: 1511-16-6 Molecular Formula: C21H24Cl2FNO2