accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1513-65-1

IUPAC Name: 2,6-difluoropyridine
Synonyms: 2,6-difluoropyridine
Molecular Weight (g/mol): 115.08
Molecular Formula: C5H3F2N
InChi Key: MBTGBRYMJKYYOE-UHFFFAOYSA-N
SMILES: FC1=CC=CC(F)=N1
Boiling Point 
  • (3)
  • (2)
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (4)
  • (5)
Percent Purity 
  • (3)
  • (2)
  • (4)
Quantity 
  • (1)
  • (4)
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (2)
  • (5)
  • (3)
  • (2)
  • (2)
  • (2)
  • (4)
  • (5)
  • (3)
  • (2)
  • (4)
  • (1)
  • (4)
  • (4)
  • (4)