Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,6-Difluoropyridine 98.0+%, TCI America™
SDP

Catalog No. D174125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D174125G 25 g
D17415G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D174125G Supplier TCI America Supplier No. D174125G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1513-65-1
Molecular Formula C5H3F2N
Molecular Weight (g/mol) 115.083
MDL Number MFCD00006227
InChI Key MBTGBRYMJKYYOE-UHFFFAOYSA-N
Synonym pyridine, 2,6-difluoro, 2,6-difluorpyridin, ccris 1720, 2,6-difluoro-pyridine, 2,6-difluoropyridin, pubchem9241, 2,6 difluoropyridine, 2, 6-difluoropyridine, 2,6 difluoro pyridine, 2,6-di-fluoropyridine
PubChem CID 73934
IUPAC Name 2,6-difluoropyridine
SMILES C1=CC(=NC(=C1)F)F

Specifications

Color Yellow
Boiling Point 125°C
Quantity 25 g
UN Number 1993
Formula Weight 115.08
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,6-Difluoropyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.