accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 15151-51-6

IUPAC Name: hydrogen 3-aminobenzoic acid chloride
Synonyms: gabaculine
Molecular Weight (g/mol): 173.6
Molecular Formula: C7H8ClNO2
InChi Key: PTTMMENRLMBXGK-UHFFFAOYSA-N
SMILES: [H+].[Cl-].NC1=CC=CC(=C1)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results