accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 151889-03-1

IUPAC Name: N-[2-(5-methoxy-2-phenyl-1H-indol-3-yl)ethyl]acetamide
Synonyms: N-[2-(5-methoxy-2-phenyl-1H-indol-3-yl)ethyl]acetamide
Molecular Weight (g/mol): 308.38
Molecular Formula: C19H20N2O2
InChi Key: OFCLARYYBGKCHN-UHFFFAOYSA-N
SMILES: COC1=CC=C2NC(=C(CCNC(C)=O)C2=C1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results