accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3CCH3OONNI

CAS RN 151978-66-4

IUPAC Name: tert-butyl 4-(4-iodophenyl)piperazine-1-carboxylate
Synonyms: tert-butyl 4-(4-iodophenyl)piperazine-1-carboxylate
Molecular Weight (g/mol): 388.25
Molecular Formula: C15H21IN2O2
InChi Key: ZOWJTZMYSLZILG-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)N1CCN(CC1)C1=CC=C(I)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results