accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 152121-47-6

IUPAC Name: 4-[4-(4-fluorophenyl)-2-(4-methanesulfinylphenyl)-1H-imidazol-5-yl]pyridine
Synonyms: 4-[5-(4-fluorophenyl)-2-(4-methanesulfinylphenyl)-3H-imidazol-4-yl]pyridine
Molecular Weight (g/mol): 377.44
Molecular Formula: C21H16FN3OS
InChi Key: CDMGBJANTYXAIV-UHFFFAOYNA-N
SMILES: CS(=O)C1=CC=C(C=C1)C1=NC(=C(N1)C1=CC=NC=C1)C1=CC=C(F)C=C1
Product Type 
  • (3)
  • (3)
Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results