CAS RN 152121-53-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
MilliporeSigma™ Calbiochem™ PD 169316
A potent, cell-permeable, reversible, competitive, and selective p38 MAP kinase inhibitor (IC50 = 89 nM).