accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1522-22-1

IUPAC Name: (3Z)-1,1,1,5,5,5-hexafluoro-4-hydroxypent-3-en-2-one
Synonyms: hexafluoroacetylacetone
Molecular Weight (g/mol): 208.06
Molecular Formula: C5H2F6O2
InChi Key: ZUSKFDORJCVUSD-UPHRSURJSA-N
SMILES: O\C(=C/C(=O)C(F)(F)F)C(F)(F)F
Boiling Point 
  • (3)
  • (2)
  • (2)
  • (3)
Molecular Weight (g/mol) 
  • (5)
  • (5)
Percent Purity 
  • (2)
  • (3)
  • (2)
  • (3)
Quantity 
  • (2)
  • (4)
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (2)
  • (2)
  • (6)
  • (3)
  • (2)
  • (2)
  • (3)
  • (5)
  • (5)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (4)
  • (4)
  • (5)