Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,1,1,5,5,5-Hexafluoroacetylacetone, 99%

Catalog No. AC218530250
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC218530250 25 g Glass bottle
AC218530050 5 g Glass bottle
AC218531000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. AC218530250 Supplier Thermo Scientific Chemicals Supplier No. 218530250
  • $266.00
    $212.65 / Each of 1

    Save $53.35

    20% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart
Add to Cart
Request Bulk or Custom Formats

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1522-22-1 | C5H2F6O2 | 208.06 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1522-22-1
Molecular Formula C5H2F6O2
Molecular Weight (g/mol) 208.06
InChI Key QAMFBRUWYYMMGJ-UHFFFAOYSA-N
Synonym hexafluoroacetylacetone, 1,1,1,5,5,5-hexafluoroacetylacetone, 1,1,1,5,5,5-hexafluoro-2,4-pentanedione, 2,4-pentanedione, 1,1,1,5,5,5-hexafluoro, hexafluoro-2,4-pentanedione, hfac, 1,1,1,5,5,5-hexafluoro-2,4-pentadione, 1,3-bis trifluoromethyl propane-1,3-dione, 1,1,1-trifluoro-3-trifluoroacetyl acetone, 2,4-dioxo-1,1,1,5,5,5-hexafluoropentane
PubChem CID 73706
IUPAC Name 1,1,1,5,5,5-hexafluoropentane-2,4-dione
SMILES C(C(=O)C(F)(F)F)C(=O)C(F)(F)F

Specifications

Color Colorless to Yellow
Density 1.4700g/mL
Boiling Point 70.0°C to 71.0°C
Flash Point 32°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Refractive Index 1.3320 to 1.3340
Linear Formula CF3COCH2COCF3
Quantity 25 g
Beilstein 01,III,3123
Specific Gravity 1.47
Solubility Information Solubility in water: immiscible
Formula Weight 208.06
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 1,1,1,5,5,5-Hexafluoroacetylacetone
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Flammable liquid and vapour.
Causes severe skin burns and eye damage.
Toxic if swallowed.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat
EINECSNumber 216-191-

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.