Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,1,1,5,5,5-Hexafluoro-2,4-pentanedione, 98+%

Catalog No. AAL0169322
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0169322 100 g
AAL0169306 5 g
AAL0169314 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL0169322 Supplier Thermo Scientific Chemicals Supplier No. L0169322
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 1522-22-1 | C5H2F6O2 | 208.059 g/mol

1,1,1,5,5,5-Hexafluoro-2,4-pentanedione acts as a chelating ligand. It is used as liquid crystal intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,1,1,5,5,5-Hexafluoro-2,4-pentanedione acts as a chelating ligand. It is used as liquid crystal intermediate.

Solubility
Not miscible in water.

Notes
Store away from oxidizing agents, reducing agents and water. Hygroscopic.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1522-22-1
Molecular Formula C5H2F6O2
Molecular Weight (g/mol) 208.059
MDL Number MFCD00000426
InChI Key QAMFBRUWYYMMGJ-UHFFFAOYSA-N
Synonym hexafluoroacetylacetone, 1,1,1,5,5,5-hexafluoroacetylacetone, 1,1,1,5,5,5-hexafluoro-2,4-pentanedione, 2,4-pentanedione, 1,1,1,5,5,5-hexafluoro, hexafluoro-2,4-pentanedione, hfac, 1,1,1,5,5,5-hexafluoro-2,4-pentadione, 1,3-bis trifluoromethyl propane-1,3-dione, 1,1,1-trifluoro-3-trifluoroacetyl acetone, 2,4-dioxo-1,1,1,5,5,5-hexafluoropentane
PubChem CID 73706
IUPAC Name 1,1,1,5,5,5-hexafluoropentane-2,4-dione
SMILES C(C(=O)C(F)(F)F)C(=O)C(F)(F)F

Specifications

Density 1.486
Boiling Point 69°C to 71°C
Flash Point 32°C (89°F)
Refractive Index 1.334
Quantity 100 g
UN Number UN2920
Beilstein 1790138
Sensitivity Hygroscopic
Solubility Information Not miscible in water.
Formula Weight 208.06
Percent Purity ≥98%
Chemical Name or Material 1,1,1,5,5,5-Hexafluoro-2,4-pentanedione
Show More Show Less
Hazard Category H226-H301-H314-H335
Hazard Statement GHS H Statement
H314-H318-H226-H302-H335
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.
Harmful if swallowed.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUIDS, FLAMMABLE, N.O.S.
EINECSNumber 216-191-0
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.