accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3CCH3H3CCH3H3CCH3CH3H3C

CAS RN 15220-85-6

IUPAC Name: 2,2,4,4,6,6,8-heptamethylnonane
Synonyms: 2-methyl-1-propene (tetramer)
Molecular Weight (g/mol): 226.45
Molecular Formula: C16H34
InChi Key: RDUBCTHNBWFPTP-UHFFFAOYSA-N
SMILES: CC(C)CC(C)(C)CC(C)(C)CC(C)(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results