accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NSNHONHNH3C

CAS RN 152239-46-8

IUPAC Name: 3-(3-methyl-1,2-thiazol-5-yl)-1-(1-methyl-1H-indol-5-yl)urea
Synonyms: 3-(3-methyl-1,2-thiazol-5-yl)-1-(1-methylindol-5-yl)urea
Molecular Weight (g/mol): 286.35
Molecular Formula: C14H14N4OS
InChi Key: USFUFHFQWXDVMH-UHFFFAOYSA-N
SMILES: CN1C=CC2=CC(NC(=O)NC3=CC(C)=NS3)=CC=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results