accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 15258-73-8

IUPAC Name: (2,6-dichlorophenyl)methanol
Synonyms: benzenemethanol, 2,6-dichloro-
Molecular Weight (g/mol): 177.02
Molecular Formula: C7H6Cl2O
InChi Key: WKKHCCZLKYKUDN-UHFFFAOYSA-N
SMILES: OCC1=C(Cl)C=CC=C1Cl
Molecular Weight (g/mol) 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results