accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClCH3H3COON(S)NClCl

CAS RN 153050-21-6

IUPAC Name: 1-{2-[(3S)-3-(3,4-dichlorophenyl)-1-{2-[3-(propan-2-yloxy)phenyl]acetyl}piperidin-3-yl]ethyl}-4-phenyl-1-azabicyclo[2.2.2]octan-1-ium chloride
Synonyms: 1-{2-[(3S)-3-(3,4-dichlorophenyl)-1-[2-(3-isopropoxyphenyl)acetyl]piperidin-3-yl]ethyl}-4-phenyl-1-azabicyclo[2.2.2]octan-1-ium chloride
Molecular Weight (g/mol): 656.13
Molecular Formula: C37H45Cl3N2O2
InChi Key: NQHFSECYQAQZBN-LSYPWIJNSA-M
SMILES: [Cl-].CC(C)OC1=CC=CC(CC(=O)N2CCC[C@@](CC[N+]34CCC(CC3)(CC4)C3=CC=CC=C3)(C2)C2=CC=C(Cl)C(Cl)=C2)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results