accessibility menu, dialog, popup

UserName

Viewing profile as user:
ON

CAS RN 1532-72-5

IUPAC Name: isoquinolin-2-ium-2-olate
Synonyms: isoquinolin-2-ium-2-olate
Molecular Weight (g/mol): 145.16
Molecular Formula: C9H7NO
InChi Key: RZIAABRFQASVSW-UHFFFAOYSA-N
SMILES: [O-][N+]1=CC=C2C=CC=CC2=C1
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)