CAS RN 1532-84-9
IUPAC Name:
isoquinolin-1-amine
Synonyms:
1-isoquinolinamine
Molecular Weight (g/mol):
144.18
Molecular Formula:
C9H8N2
InChi Key:
OSILBMSORKFRTB-UHFFFAOYSA-N
SMILES:
NC1=C2C=CC=CC2=CC=N1
1
–
1
of
1
results
1
1-Aminoisoquinoline 98.0+%, TCI America™
CAS: 1532-84-9 Molecular Formula: C9H8N2 Molecular Weight (g/mol): 144.18 MDL Number: MFCD00024137 InChI Key: OSILBMSORKFRTB-UHFFFAOYSA-N PubChem CID: 73742 IUPAC Name: isoquinolin-1-amine SMILES: NC1=C2C=CC=CC2=CC=N1