accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OFFF

CAS RN 1533-03-5

IUPAC Name: 1-[3-(trifluoromethyl)phenyl]propan-1-one
Synonyms: 1-[3-(trifluoromethyl)phenyl]propan-1-one
Molecular Weight (g/mol): 202.18
Molecular Formula: C10H9F3O
InChi Key: UBTPNKXHYILGJU-UHFFFAOYSA-N
SMILES: CCC(=O)C1=CC=CC(=C1)C(F)(F)F
Boiling Point 
  • (2)
  • (1)
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)