accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3ONHOOOHOHOHHOOH3C(E/Z)

CAS RN 153409-16-6

IUPAC Name: N-[(3-methoxy-4-{[3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}phenyl)methyl]-8-methylnon-6-enamide
Synonyms: N-[(3-methoxy-4-{[3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}phenyl)methyl]-8-methylnon-6-enamide
Molecular Weight (g/mol): 467.56
Molecular Formula: C24H37NO8
InChi Key: HEYWYCJIIXVRPS-UHFFFAOYNA-N
SMILES: COC1=C(OC2OC(CO)C(O)C(O)C2O)C=CC(CNC(=O)CCCCC=CC(C)C)=C1
Grade 
  • (1)
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)