accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 153436-38-5

IUPAC Name: 4,6-dichloro-3-[2-(phenylcarbamoyl)eth-1-en-1-yl]-1H-indole-2-carboxylic acid sodium
Synonyms: 4,6-dichloro-3-[2-(phenylcarbamoyl)eth-1-en-1-yl]-1H-indole-2-carboxylic acid sodium
Molecular Weight (g/mol): 398.19
Molecular Formula: C18H12Cl2N2NaO3
InChi Key: OBPMZHMRJJJUBC-UHFFFAOYSA-N
SMILES: [Na].OC(=O)C1=C(C=CC(=O)NC2=CC=CC=C2)C2=C(Cl)C=C(Cl)C=C2N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results