CAS RN 153559-76-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Tocris Bioscience™ LG 100268
Potent and selective RXR agonist; also activates RXR/PPARγ heterodimers