accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 15356-62-4

IUPAC Name: 2-{[5-methyl-2-(propan-2-yl)cyclohexyl]oxy}acetyl chloride
Synonyms: 2-[(2-isopropyl-5-methylcyclohexyl)oxy]acetyl chloride
Molecular Weight (g/mol): 232.75
Molecular Formula: C12H21ClO2
InChi Key: VNMCKLVHDJADEB-UHFFFAOYNA-N
SMILES: CC(C)C1CCC(C)CC1OCC(Cl)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results