accessibility menu, dialog, popup

UserName

Viewing profile as user:
NNBr

CAS RN 1537218-76-0

IUPAC Name: 3-bromo-9-[4-(9H-carbazol-9-yl)phenyl]-9H-carbazole
Synonyms: 3-bromo-9-[4-(carbazol-9-yl)phenyl]carbazole
Molecular Weight (g/mol): 487.4
Molecular Formula: C30H19BrN2
InChi Key: NEYCIRFFXNWOAH-UHFFFAOYSA-N
SMILES: BrC1=CC2=C(C=C1)N(C1=C2C=CC=C1)C1=CC=C(C=C1)N1C2=C(C=CC=C2)C2=C1C=CC=C2
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results