CAS RN 1542705-92-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
R&D Systems™ CB 5083
A Tocris product, part of R&D Systems. Potent p97 ATPase inhibitor