accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOOHNONHOCl

CAS RN 15474-96-1

IUPAC Name: 2-[2-(2-chloroacetamido)acetamido]acetic acid
Synonyms: [2-(2-chloroacetamido)acetamido]acetic acid
Molecular Weight (g/mol): 208.6
Molecular Formula: C6H9ClN2O4
InChi Key: NCOXSGKCQMMTRX-UHFFFAOYSA-N
SMILES: OC(=O)CNC(=O)CNC(=O)CCl
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results