accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 154748-49-9

IUPAC Name: tert-butyl N-(3-oxocyclobutyl)carbamate
Synonyms: tert-butyl N-(3-oxocyclobutyl)carbamate
Molecular Weight (g/mol): 185.22
Molecular Formula: C9H15NO3
InChi Key: FNHPTFKSPUTESA-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)NC1CC(=O)C1
Molecular Weight (g/mol) 
  • (2)
  • (1)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)