Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

tert-Butyl (3-oxocyclobutyl)carbamate, 95%

Catalog No. AC446590050
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC446590050 5 g
AC446590010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC446590050 Supplier Thermo Scientific Chemicals Supplier No. 446590050
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 154748-49-9 | C9H15NO3 | 185.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 154748-49-9
Molecular Formula C9H15NO3
Molecular Weight (g/mol) 185.22
InChI Key FNHPTFKSPUTESA-UHFFFAOYSA-N
Synonym tert-butyl 3-oxocyclobutylcarbamate, tert-butyl 3-oxocyclobutyl carbamate, tert-butyl n-3-oxocyclobutyl carbamate, 3-oxo-cyclobutyl-carbamic acid tert-butyl ester, 3-boc-amino cyclobutanone, 2-methyl-2-propanyl 3-oxocyclobutyl carbamate, 3-tert-butyloxycarbonylamino cyclobutan-1-one, 3-oxocyclobutylcarbamic acid, 1,1-dimethylethyl ester, carbamic acid, 3-oxocyclobutyl-, 1,1-dimethylethyl ester, carbamic acid, n-3-oxocyclobutyl-, 1,1-dimethylethyl ester
PubChem CID 23374584
IUPAC Name tert-butyl N-(3-oxocyclobutyl)carbamate
SMILES CC(C)(C)OC(=O)NC1CC(=O)C1

Specifications

Melting Point 122°C to 124°C
Assay Percent Range 94% min. (GC)
Quantity 5 g
Formula Weight 185.22
Percent Purity 95%
Chemical Name or Material tert-Butyl (3-oxocyclobutyl)carbamate
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.