Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-(tert-Butoxycarbonylamino)-1-cyclobutanone 98.0+%, TCI America™
SDP

Catalog No. B4043200MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B4043200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B4043200MG Supplier TCI America Supplier No. B4043200MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

A protected amino-cyclobutanone reagent used for chemical synthesis.

Chemical Identifiers

CAS 154748-49-9
Molecular Formula C9H15NO3
Molecular Weight (g/mol) 185.223
MDL Number MFCD09751872
InChI Key FNHPTFKSPUTESA-UHFFFAOYSA-N
Synonym tert-butyl 3-oxocyclobutylcarbamate, tert-butyl 3-oxocyclobutyl carbamate, tert-butyl n-3-oxocyclobutyl carbamate, 3-oxo-cyclobutyl-carbamic acid tert-butyl ester, 3-boc-amino cyclobutanone, 2-methyl-2-propanyl 3-oxocyclobutyl carbamate, 3-tert-butyloxycarbonylamino cyclobutan-1-one, 3-oxocyclobutylcarbamic acid, 1,1-dimethylethyl ester, carbamic acid, 3-oxocyclobutyl-, 1,1-dimethylethyl ester, carbamic acid, n-3-oxocyclobutyl-, 1,1-dimethylethyl ester
PubChem CID 23374584
IUPAC Name tert-butyl N-(3-oxocyclobutyl)carbamate
SMILES CC(C)(C)OC(=O)NC1CC(=O)C1

Specifications

Melting Point 123°C
Quantity 200 mg
Formula Weight 185.22
Percent Purity ≥98.0% (GC)
Grade Research
Physical Form Crystalline Powder
Chemical Name or Material 3-(tert-Butoxycarbonylamino)-1-cyclobutanone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.