accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 155059-42-0

IUPAC Name: 8-methyl-8-azabicyclo[3.2.1]octan-3-yl 2-(4-chlorophenoxy)butanoate; but-2-enedioic acid
Synonyms: 8-methyl-8-azabicyclo[3.2.1]octan-3-yl 2-(4-chlorophenoxy)butanoate; butenedioic acid
Molecular Weight (g/mol): 453.92
Molecular Formula: C22H28ClNO7
InChi Key: BHXGTFUQDGMXHA-UHFFFAOYNA-N
SMILES: OC(=O)C=CC(O)=O.CCC(OC1=CC=C(Cl)C=C1)C(=O)OC1CC2CCC(C1)N2C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results